missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(3-pyridylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15480756
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449132-32-4
Molecular Formula C12H10BFN2O3
Molecular Weight (g/mol) 260.031
MDL Number MFCD20265175
InChI Key LWVDXBKALYJJIU-UHFFFAOYSA-N
Synonym 2-fluoro-5-3-pyridylcarbamoyl benzeneboronic acid, 2-fluoro-5-pyridin-3-yl carbamoyl phenylboronic acid, 2-fluoro-5-3-pyridylcarbamoyl phenylboronic acid
PubChem CID 73995628
IUPAC Name [2-fluoro-5-(pyridin-3-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)NC2=CN=CC=C2)F)(O)O

Specifications

Quantity 250 mg
Formula Weight 260.03
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(3-pyridylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'