missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™
£100.00 - £302.00
Chemical Identifiers
CAS | 944317-66-2 |
---|---|
Molecular Formula | C15H20BFO4 |
Molecular Weight (g/mol) | 294.13 |
MDL Number | MFCD12026092 |
InChI Key | LTWSFTBHFDGCTE-UHFFFAOYSA-N |
PubChem CID | 46739456 |
IUPAC Name | methyl 2-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate |
SMILES | COC(=O)CC1=CC(B2OC(C)(C)C(C)(C)O2)=C(F)C=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
944317-66-2 | |
294.13 | |
LTWSFTBHFDGCTE-UHFFFAOYSA-N | |
methyl 2-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate |
C15H20BFO4 | |
MFCD12026092 | |
46739456 | |
COC(=O)CC1=CC(B2OC(C)(C)C(C)(C)O2)=C(F)C=C1 |
Specifications
944317-66-2 | |
MFCD12026092 | |
LTWSFTBHFDGCTE-UHFFFAOYSA-N | |
methyl 2-[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate | |
46739456 | |
96% | |
2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester |
C15H20BFO4 | |
methyl 2-4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 2-fluoro-5-methoxycarbonylmethyl phenylboronic acid, pinacol ester, methyl 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 2-fluoro-5-methoxycarbonylmethyl phenylboronic acid,pinacol ester, 2-fluoro-5-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-4-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, methyl2-4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetic acid methyl ester | |
COC(=O)CC1=CC(B2OC(C)(C)C(C)(C)O2)=C(F)C=C1 | |
294.13 | |
294.13 | |
Solid |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures