missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11342559
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-5-(trifluoromethoxy)benzaldehyde is used as a precursor for organic synthesis. It is also used as a pharmaceutical intermediate.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents, strong reducing agents, strong acids, ammonia and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886497-81-0
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
MDL Number MFCD04115886
InChI Key FMDGCHMPSCBYFX-UHFFFAOYSA-N
PubChem CID 2783324
IUPAC Name 2-fluoro-5-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1OC(F)(F)F)C=O)F

Specifications

Refractive Index 1.443
Quantity 1 g
Sensitivity Air Sensitive
Synonym 2-fluoro-5-trifluoromethoxy benzaldehyde, 2-formyl-4-trifluoromethoxy fluorobenzene, benzaldehyde,2-fluoro-5-trifluoromethoxy, pubchem4231, 2-fluoro-5-trifluoromethoxy-benzaldehyde, benzaldehyde, 2-fluoro-5-trifluoromethoxy
Solubility Information Slightly miscible with water.
Formula Weight 208.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.