Learn More
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific™
£42.10 - £159.72
Chemical Identifiers
CAS | 69922-27-6 |
---|---|
Molecular Formula | C8H3F4NO |
Molecular Weight (g/mol) | 205.11 |
MDL Number | MFCD00673069 |
InChI Key | NAIKHCBDZGSGHH-UHFFFAOYSA-N |
PubChem CID | 2733380 |
IUPAC Name | 1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene |
SMILES | FC1=CC=C(C=C1N=C=O)C(F)(F)F |
Description
Chemical Identifiers
69922-27-6 | |
205.11 | |
NAIKHCBDZGSGHH-UHFFFAOYSA-N | |
1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene |
C8H3F4NO | |
MFCD00673069 | |
2733380 | |
FC1=CC=C(C=C1N=C=O)C(F)(F)F |
Specifications
69922-27-6 | |
100.0 | |
53°C (2.5 mmHg) | |
97% | |
FC6H3(CF3)NCO | |
MFCD00673069 | |
NAIKHCBDZGSGHH-UHFFFAOYSA-N | |
1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene | |
2733380 | |
97% | |
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97% |
96.0 | |
Colorless | |
Authentic | |
C8H3F4NO | |
1.4540 to 1.456 | |
2-fluoro-5-trifluoromethyl phenyl isocyanate, 1-fluoro-2-isocyanato-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl phenylisocyanate, 2-fluoro-5-trifluoromethylphenyl isocyanate, benzene,1-fluoro-2-isocyanato-4-trifluoromethyl, benzene, 1-fluoro-2-isocyanato-4-trifluoromethyl, acmc-20anuz, pubchem12480, 2-fluoro-5-trifluoromethylphenylisocyanate, 2-fluoro-5-trifluoromethyl phenyl isocyan | |
FC1=CC=C(C=C1N=C=O)C(F)(F)F | |
205.11 | |
205.11 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'tsca' : TSCA
RUO – Research Use Only