missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzyl bromide, 98%, Thermo Scientific Chemicals

Product Code. 11342139
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500912-17-4
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.053
MDL Number MFCD08688579
InChI Key PVKFBCBTHHTDEX-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxybenzyl bromide, 2-bromomethyl-1-fluoro-3-methoxybenzene, 2-bromomethyl-3-fluoroanisole, 2-fluoro-6-methoxy-benzyl bromide, 2-bromomethyl-3-fluorophenyl methyl ether, benzene,2-bromomethyl-1-fluoro-3-methoxy, 2-bromomethyl-1-fluoranyl-3-methoxy-benzene
PubChem CID 2779322
IUPAC Name 2-(bromomethyl)-1-fluoro-3-methoxybenzene
SMILES COC1=C(C(=CC=C1)F)CBr

Specifications

Melting Point 18°C to 21°C
Refractive Index 1.557
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 219.05
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-methoxybenzyl bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'