missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-nitrotoluene, 98%, Thermo Scientific™

Product Code. 11314536
Click to view available options
Quantity:
5 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-10-8
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00007197
InChI Key GXPIVRKDWZKIKZ-UHFFFAOYSA-N
Synonym 2-fluoro-6-nitrotoluene, 1-fluor-2-methyl-3-nitrobenzene, 2-nitro-6-fluorotoluene, 6-fluoro-2-nitrotoluene, benzene, 1-fluoro-2-methyl-3-nitro, 3-fluoro-2-methyl-1-nitrobenzene, 1-fluor-2-methyl-3-nitrobenzol, pubchem1594, acmc-1bc1k
PubChem CID 69854
IUPAC Name 1-fluoro-2-methyl-3-nitrobenzene
SMILES CC1=C(F)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 6°C to 8°C
Density 1.277
Boiling Point 208°C to 210°C
Flash Point 88°C (190°F)
Refractive Index 1.524
Quantity 100 g
UN Number UN2810
Beilstein 2361978
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-nitrotoluene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'