missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-phenoxybenzaldehyde, 96%, Thermo Scientific™

Product Code. 11444870
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-68-4
Molecular Formula C13H9FO2
Molecular Weight (g/mol) 216.211
MDL Number MFCD03428585
InChI Key AUPPVEMCDSSXCW-UHFFFAOYSA-N
Synonym 2-fluoro-6-phenoxy-benzaldehyde
PubChem CID 24721181
IUPAC Name 2-fluoro-6-phenoxybenzaldehyde
SMILES C1=CC=C(C=C1)OC2=C(C(=CC=C2)F)C=O

Specifications

Melting Point 36°C to 39°C
Quantity 1 g
Formula Weight 216.21
Percent Purity 96%
Chemical Name or Material 2-Fluoro-6-phenoxybenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'