missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific™
£43.70 - £158.00
Chemical Identifiers
CAS | 313545-34-5 |
---|---|
Molecular Formula | C7H5BF4O2 |
Molecular Weight (g/mol) | 207.919 |
MDL Number | MFCD07781239 |
InChI Key | TXBRJTKFAYXKMV-UHFFFAOYSA-N |
Synonym | 2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenylboron, 2-fluoro-6-trifluoromethylphenyl boronic acid |
PubChem CID | 16217168 |
IUPAC Name | [2-fluoro-6-(trifluoromethyl)phenyl]boronic acid |
SMILES | B(C1=C(C=CC=C1F)C(F)(F)F)(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
313545-34-5 | |
207.919 | |
TXBRJTKFAYXKMV-UHFFFAOYSA-N | |
16217168 | |
B(C1=C(C=CC=C1F)C(F)(F)F)(O)O |
C7H5BF4O2 | |
MFCD07781239 | |
2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenylboron, 2-fluoro-6-trifluoromethylphenyl boronic acid | |
[2-fluoro-6-(trifluoromethyl)phenyl]boronic acid |
Specifications
313545-34-5 | |
C7H5BF4O2 | |
2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenylboron, 2-fluoro-6-trifluoromethylphenyl boronic acid | |
B(C1=C(C=CC=C1F)C(F)(F)F)(O)O | |
207.919 | |
207.92 | |
Solid |
115°C to 119°C | |
MFCD07781239 | |
TXBRJTKFAYXKMV-UHFFFAOYSA-N | |
[2-fluoro-6-(trifluoromethyl)phenyl]boronic acid | |
16217168 | |
98% | |
2-Fluoro-6-(trifluoromethyl)benzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures