missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2'-Fluoro-Bz-A Phosphoramidite, standard grade, 20-400 finish bottle

Product Code. 17964272 Shop All Thermo Scientific Products
Click to view available options
Quantity:
0.25 g
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17964272

Brand: Thermo Scientific™ 27160109

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2'-Fluoro-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-Fluoro protection, for use in oligonucleotide synthesis.

2'-Fluoro-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-Fluoro protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, the highly electronegative fluorine (2'-Fluoro) modification adopts a more thermodynamically stable A-form helix on hybridization to its target. Showing improved affinity and some nuclease resistance (compared to 2'-OH), the 2'-F modification has been proven useful in antisense, siRNA, and aptamer applications. This product is supplied in a 30-mL amber bottle with a 20-400 finish neck style, which is compatible with most Expedite synthesizers.

TRUSTED_SUSTAINABILITY

Specifications

31P NMR Purity ≥98%
Format Amber bottle (20-400 finish neck style)
For Use With (Equipment) Expedite Synthesizers
Description Base: A
Base Protection: Bz
Molecular Weight (g/mol) 875.92 g/mol
Product Type 2in-Fluoro-Bz-A CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Adenosine (A)
Reagent Features 2in-Fluoro modified
Shipping Condition Room Temperature
Purity >98% (HPLC)
Quantity 0.25 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type RNA
For Use With (Application) Oligonucleotide synthesis
Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-N6-benzoyl-2'-fluoroadenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular Formula C47H51FN7O7P
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.