missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-L-phenylalanine, 99+%, Thermo Scientific Chemicals

Product Code. 15477725
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19883-78-4
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.18
MDL Number MFCD00066448
InChI Key NYCRCTMDYITATC-QMMMGPOBSA-N
Synonym 2-fluoro-l-phenylalanine, h-phe 2-f-oh, l-2-fluorophenylalanine, 2s-2-amino-3-2-fluorophenyl propanoic acid, l-2-f-phe-oh, o-fluoro-l-phenylalanine, 2-fluoro-l-phe, phenylalanine, 2-fluoro, s-2-amino-3-2-fluorophenyl propanoic acid, l-2-fluorophe
PubChem CID 716319
IUPAC Name (2S)-2-amino-3-(2-fluorophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC=C1F)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 183.18
Percent Purity ≥99%
Chemical Name or Material 2-Fluoro-L-phenylalanine, sum of enantiomers

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'