Learn More
2-Fluoro-m-xylene, 98%, Thermo Scientific Chemicals
£43.00 - £158.00
Chemical Identifiers
CAS | 443-88-9 |
---|---|
Molecular Formula | C8H9F |
Molecular Weight (g/mol) | 124.158 |
MDL Number | MFCD00039215 |
InChI Key | JTAUTNBVFDTYTI-UHFFFAOYSA-N |
Synonym | 2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene |
PubChem CID | 123065 |
IUPAC Name | 2-fluoro-1,3-dimethylbenzene |
SMILES | CC1=C(C(=CC=C1)C)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11468623
|
Thermo Scientific Alfa Aesar
A10424.06 |
5 g |
£43.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478623
|
Thermo Scientific Alfa Aesar
A10424.14 |
25 g |
£158.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
Chemical Identifiers
443-88-9 | |
124.158 | |
JTAUTNBVFDTYTI-UHFFFAOYSA-N | |
123065 | |
CC1=C(C(=CC=C1)C)F |
C8H9F | |
MFCD00039215 | |
2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene | |
2-fluoro-1,3-dimethylbenzene |
Specifications
443-88-9 | |
147°C to 148°C | |
1.479 | |
MFCD00039215 | |
1857413 | |
Not miscible or difficult to mix in water. | |
CC1=C(C(=CC=C1)C)F | |
124.158 | |
124.16 | |
2-Fluoro-m-xylene |
0.988 | |
Aromatic | |
C8H9F | |
UN1993 | |
2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene | |
JTAUTNBVFDTYTI-UHFFFAOYSA-N | |
2-fluoro-1,3-dimethylbenzene | |
123065 | |
98% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 207-144-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only