missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzoic acid, 98+%, Thermo Scientific Chemicals

 

£39.30 - £158.79

Chemical Identifiers

CAS 445-29-4
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD00002405
InChI Key NSTREUWFTAOOKS-UHFFFAOYSA-N
Synonym o-fluorobenzoic acid, benzoic acid, 2-fluoro, ortho-fluorobenzoic acid, o-fluorbenzoesaeure, benzoic acid, o-fluoro, fluorobenzoic acid, unii-64uz32koo4, o-fluorbenzoesaeure german, ofba, 2-fluoro-benzoic acid
PubChem CID 9935
ChEBI CHEBI:19577
IUPAC Name 2-fluorobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11434493
SDS View Documents
Thermo Scientific Alfa Aesar
A11015.22
100 g
£39.30
100g
Please to purchase this item. Need a web account? Register with us today!
11444493
SDS View Documents
Thermo Scientific Alfa Aesar
A11015.36
500 g
£158.79
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluorobenzoic acid, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.