missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11444493
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-29-4
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD00002405
InChI Key NSTREUWFTAOOKS-UHFFFAOYSA-N
Synonym o-fluorobenzoic acid, benzoic acid, 2-fluoro, ortho-fluorobenzoic acid, o-fluorbenzoesaeure, benzoic acid, o-fluoro, fluorobenzoic acid, unii-64uz32koo4, o-fluorbenzoesaeure german, ofba, 2-fluoro-benzoic acid
PubChem CID 9935
ChEBI CHEBI:19577
IUPAC Name 2-fluorobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)F

Specifications

Melting Point 122°C to 125°C
Density 1.46
Quantity 500 g
Beilstein 971265
Solubility Information Slightly soluble in water.
Formula Weight 140.11
Percent Purity ≥98%
Chemical Name or Material 2-Fluorobenzoic acid
compliance-icons
Product Identifier
  • 2-Fluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'