missing translation for 'onlineSavingsMsg'
Learn More
2-Fluorobenzoyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 11473117

2-Fluorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11473117
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11473117

Brand: Thermo Scientific Alfa Aesar A16096.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-52-2
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000656
InChI Key RAAGZOYMEQDCTD-UHFFFAOYSA-N
Synonym o-fluorobenzoyl chloride, benzoyl chloride, 2-fluoro, benzoyl chloride, o-fluoro, 2-fluorobenzoylchloride, 2-fluoro benzoyl chloride, unii-z4h017n76s, ccris 5914, fluorobenzoyl chloride, benzoyl chloride, fluoro, 2-fluorobenzoic acid chloride
PubChem CID 9808
IUPAC Name 2-fluorobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)F

Specifications

Melting Point 4°C to 6°C
Boiling Point 207°C
Flash Point 82°C (179°F)
Odor Pungent
Quantity 25 g
UN Number UN3265
Beilstein 636864
Formula Weight 158.56
Percent Purity 97%
Chemical Name or Material 2-Fluorobenzoyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluorobenzoyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.