missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl bromide, 98%, Thermo Scientific Chemicals

Product Code. 10092540
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-48-0
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.03
MDL Number MFCD00000324
InChI Key FFWQLZFIMNTUCZ-UHFFFAOYSA-N
Synonym 2-fluorobenzyl bromide, 1-bromomethyl-2-fluorobenzene, 2-fluorobenzylbromide, o-fluorobenzyl bromide, alpha-bromo-o-fluorotoluene, alpha-bromo-2-fluorotoluene, 2-fluoro-benzyl bromide, 1-bromomethyl-2-fluoro-benzene, benzene, 1-bromomethyl-2-fluoro, 2-fluoro-benzylbromide
PubChem CID 67968
IUPAC Name 1-(bromomethyl)-2-fluorobenzene
SMILES C1=CC=C(C(=C1)CBr)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5670g/mL
Boiling Point 84°C to 85°C (15.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H4CH2Br
Refractive Index 1.5495 to 1.5515
Quantity 5 g
Beilstein 05, II, 238
Specific Gravity 1.567
Formula Weight 189.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzyl bromide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'