missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorobiphenyl, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
10 g
25 g
Description
2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.
Solubility
Soluble in alcohol, ether. Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.
Solubility
Soluble in alcohol, ether. Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 72°C to 76°C |
Density | 1.245 |
Boiling Point | 248°C |
Quantity | 5 g |
Beilstein | 2043175 |
Solubility Information | Soluble in alcohol,ether. Insoluble in water. |
IUPAC Name | 1-fluoro-2-phenylbenzene |
Formula Weight | 172.2 |
Percent Purity | 98% |
Chemical Name or Material | 2-Fluorobiphenyl |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'