missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorocinnamic acid, 98%, Thermo Scientific™

Product Code. 10401831
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 451-69-4
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.15
MDL Number MFCD00004370
InChI Key IOUDZAFBPDDAMK-AATRIKPKSA-N
Synonym 2-fluorocinnamic acid, o-fluorocinnamic acid, cinnamic acid, o-fluoro, 2-propenoic acid, 3-2-fluorophenyl, 2e-3-2-fluorophenyl prop-2-enoic acid, 3-2-fluorophenyl acrylic acid, 3-2-fluoro-phenyl-acrylic acid, 2e-3-2-fluorophenyl acrylic acid, 3-2-fluorophenyl-2-propenoic acid, trans-2-fluorocinnamic acid
PubChem CID 735833
IUPAC Name (E)-3-(2-fluorophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 179°C to 183°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H4CH=CHCO2H
Quantity 25 g
Beilstein 09, 594
Formula Weight 166.15
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2-Fluorocinnamic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluorocinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'