Learn More
2-Fluorocinnamic acid, 98%, Thermo Scientific™
£14.70 - £112.00
Chemical Identifiers
CAS | 451-69-4 |
---|---|
Molecular Formula | C9H7FO2 |
Molecular Weight (g/mol) | 166.15 |
MDL Number | MFCD00004370 |
InChI Key | IOUDZAFBPDDAMK-AATRIKPKSA-N |
Synonym | 2-fluorocinnamic acid, o-fluorocinnamic acid, cinnamic acid, o-fluoro, 2-propenoic acid, 3-2-fluorophenyl, 2e-3-2-fluorophenyl prop-2-enoic acid, 3-2-fluorophenyl acrylic acid, 3-2-fluoro-phenyl-acrylic acid, 2e-3-2-fluorophenyl acrylic acid, 3-2-fluorophenyl-2-propenoic acid, trans-2-fluorocinnamic acid |
PubChem CID | 735833 |
IUPAC Name | (E)-3-(2-fluorophenyl)prop-2-enoic acid |
SMILES | OC(=O)\C=C\C1=CC=CC=C1F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10300554
|
Thermo Scientific Acros
207100010 |
1 g | Glass bottle | ||||||
10606792
|
Thermo Scientific Acros
207100050 |
5 g | Glass bottle | ||||||
10401831
|
Thermo Scientific Acros
207100250 |
25 g | Glass bottle |
£112.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
451-69-4 | |
166.15 | |
IOUDZAFBPDDAMK-AATRIKPKSA-N | |
735833 | |
OC(=O)\C=C\C1=CC=CC=C1F |
C9H7FO2 | |
MFCD00004370 | |
2-fluorocinnamic acid, o-fluorocinnamic acid, cinnamic acid, o-fluoro, 2-propenoic acid, 3-2-fluorophenyl, 2e-3-2-fluorophenyl prop-2-enoic acid, 3-2-fluorophenyl acrylic acid, 3-2-fluoro-phenyl-acrylic acid, 2e-3-2-fluorophenyl acrylic acid, 3-2-fluorophenyl-2-propenoic acid, trans-2-fluorocinnamic acid | |
(E)-3-(2-fluorophenyl)prop-2-enoic acid |
Specifications
451-69-4 | |
100.0 | |
White | |
98% | |
FC6H4CH=CHCO2H | |
09, 594 | |
IOUDZAFBPDDAMK-AATRIKPKSA-N | |
(E)-3-(2-fluorophenyl)prop-2-enoic acid | |
735833 | |
98% | |
2-Fluorocinnamic acid, 98% |
97.5 | |
179°C to 183°C | |
Authentic | |
C9H7FO2 | |
MFCD00004370 | |
2-fluorocinnamic acid, o-fluorocinnamic acid, cinnamic acid, o-fluoro, 2-propenoic acid, 3-2-fluorophenyl, 2e-3-2-fluorophenyl prop-2-enoic acid, 3-2-fluorophenyl acrylic acid, 3-2-fluoro-phenyl-acrylic acid, 2e-3-2-fluorophenyl acrylic acid, 3-2-fluorophenyl-2-propenoic acid, trans-2-fluorocinnamic acid | |
OC(=O)\C=C\C1=CC=CC=C1F | |
166.15 | |
166.15 | |
Crystalline Powder or Flakes |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-195-3
missing translation for 'rtecsNumber' : GD9081000
missing translation for 'tsca' : TSCA
- 2-Fluorocinnamic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only