missing translation for 'onlineSavingsMsg'
Learn More

(2-Fluorophenyl)methanesulfonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10408904
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 24974-71-8
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.63
MDL Number MFCD04117455
InChI Key CJDWSUDXKHLMCY-UHFFFAOYSA-N
Synonym 2-fluorophenyl methanesulfonyl chloride, 2-fluoro-phenyl-methanesulfonyl chloride, 2-fluorobenzylsulfonyl chloride, 2-fluorophenyl methanesulphonyl chloride, chloro 2-fluorophenyl methyl sulfone, alpha-chlorosulphonyl-2-fluorotoluene, benzenemethanesulfonyl chloride, 2-fluoro, 2-fluorophenyl-methanesulfonyl chloride, pubchem10055
PubChem CID 573052
IUPAC Name (2-fluorophenyl)methanesulfonyl chloride
SMILES FC1=CC=CC=C1CS(Cl)(=O)=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 208.64
Percent Purity 95%
Chemical Name or Material (2-Fluorophenyl)methanesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'