missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorothiophenol, 97%, Thermo Scientific Chemicals

Product Code. 11481577
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2557-78-0
Molecular Formula C6H4FS
Molecular Weight (g/mol) 127.16
MDL Number MFCD00041419
InChI Key WJTZZPVVTSDNJJ-UHFFFAOYSA-M
Synonym 2-fluorothiophenol, o-fluorothiophenol, o-fluorobenzenethiol, 2-fluorobenzene-1-thiol, fluorothiophenol, o-fluoro thiophenol, 2-fluoro-thiophenol, 2-fluoro-benzenethiol, 2-fluorobenzene thiol, pubchem5428
PubChem CID 520216
SMILES FC1=CC=CC=C1[S-]

Specifications

Density 1.203
Boiling Point 164°C
Flash Point 56°C (133°F)
Odor Stench
Refractive Index 1.555
Quantity 1 g
UN Number UN3071
Beilstein 2039768
Sensitivity Air Sensitive
IUPAC Name 2-fluorobenzenethiol
Formula Weight 128.17
Percent Purity 97%
Chemical Name or Material 2-Fluorothiophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'