missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Fmoc-amino)benzoic acid, 98%, Thermo Scientific Chemicals
£47.70 - £214.00
Chemical Identifiers
CAS | 150256-42-1 |
---|---|
Molecular Formula | C22H17NO4 |
Molecular Weight (g/mol) | 359.381 |
MDL Number | MFCD00235883 |
InChI Key | CNAVPEPPAVHHKN-UHFFFAOYSA-N |
Synonym | fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid |
PubChem CID | 978386 |
IUPAC Name | 2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid |
SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15430169
|
Thermo Scientific Alfa Aesar
H63459.06 |
5 g |
£47.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15440169
|
Thermo Scientific Alfa Aesar
H63459.14 |
25 g |
£214.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field
Solubility
Soluble in water or 1% acetic acid.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
150256-42-1 | |
359.381 | |
CNAVPEPPAVHHKN-UHFFFAOYSA-N | |
978386 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O |
C22H17NO4 | |
MFCD00235883 | |
fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid | |
2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid |
Specifications
150256-42-1 | |
White | |
MFCD00235883 | |
fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid | |
CNAVPEPPAVHHKN-UHFFFAOYSA-N | |
2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid | |
978386 | |
98% | |
2-(Fmoc-amino)benzoic acid |
216°C to 218°C (decomposition) | |
C22H17NO4 | |
7659726 | |
Soluble in water or 1% acetic acid. | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O | |
359.381 | |
359.37 | |
Powder |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only