missing translation for 'onlineSavingsMsg'
Learn More

2-Furaldehyde, 99%, Thermo Scientific Chemicals

£21.91 - £423.11

Chemical Identifiers

CAS 98-01-1
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003229
InChI Key HYBBIBNJHNGZAN-UHFFFAOYSA-N
Synonym furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
PubChem CID 7362
ChEBI CHEBI:34768
IUPAC Name furan-2-carbaldehyde
SMILES O=CC1=CC=CO1
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10307940
View Documents Promotion Details
Thermo Scientific Acros
181100250
25 mL Glass bottle
£21.91
25mL
Please to purchase this item. Need a web account? Register with us today!
10686382
View Documents
Thermo Scientific Acros
181102500
250 mL Glass bottle
£23.12
250mL
Please to purchase this item. Need a web account? Register with us today!
10449500
View Documents
Thermo Scientific Acros
181100010
1 L Glass bottle
£59.60
1 L
Please to purchase this item. Need a web account? Register with us today!
10074053
View Documents
Thermo Scientific Acros
181100025
2.5 L Glass bottle
£131.66
2.50 L
Please to purchase this item. Need a web account? Register with us today!
13498418
View Documents
Thermo Scientific Acros
181100100
10 L Metal drum
£423.11
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-01-1
96.09
HYBBIBNJHNGZAN-UHFFFAOYSA-N
7362
furan-2-carbaldehyde
C5H4O2
MFCD00003229
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
CHEBI:34768
O=CC1=CC=CO1

Specifications

0.02 eq/L max.
98.5
-37°C
1.1600g/mL
60°C
98.5% min. (GC)
1.5250 to 1.527
0.5% max.
1.16
Solubility in water: 83g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene
O=CC1=CC=CO1
96.09
CHEBI:34768
96.08
Liquid
98-01-1
100.0
Amber to Brown
159°C to 161°C
Authentic
C5H4O2
MFCD00003229
15,4333
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
HYBBIBNJHNGZAN-UHFFFAOYSA-N
furan-2-carbaldehyde
7362
1.49 mPa.s (25°C)
99%
2-Furaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Sus

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to f

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-627-7

Recommended Storage : This product darkens on contact with air

Product Identifier
  • 2-Furaldehyde

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only