missing translation for 'onlineSavingsMsg'
Learn More

2-Furamide, 97%, Thermo Scientific Chemicals

Product Code. 11426067
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-38-1
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00236147
InChI Key TVFIYRKPCACCNL-UHFFFAOYSA-N
Synonym 2-furamide, furamide, 2-furancarboxamide, furfurylamide, 2-furoamide, furoylamide, unii-8ig4632c1y, furancarboxamide, acmc-20anu6
PubChem CID 69108
IUPAC Name furan-2-carboxamide
SMILES C1=COC(=C1)C(=O)N

Specifications

Melting Point 139°C to 142°C
Quantity 25 g
Formula Weight 111.1
Percent Purity 97%
Chemical Name or Material 2-Furamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'