Learn More
2-Furonitrile, 99%, Thermo Scientific Chemicals
£81.60 - £276.00
Chemical Identifiers
CAS | 617-90-3 |
---|---|
Molecular Formula | C5H3NO |
Molecular Weight (g/mol) | 93.09 |
MDL Number | MFCD00003223 |
InChI Key | YXDXXGXWFJCXEB-UHFFFAOYSA-N |
Synonym | 2-furonitrile, 2-cyanofuran, 2-furancarbonitrile, 2-furyl cyanide, unii-2lrk86h722, .alpha.-furyl cyanide, pubchem6961, acmc-209mwb, 2-cyanofuran;2-furancarbonitrile |
PubChem CID | 69245 |
IUPAC Name | furan-2-carbonitrile |
SMILES | N#CC1=CC=CO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10205700
|
Thermo Scientific Acros
170430050 |
5 g | Glass bottle |
£81.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10645802
|
Thermo Scientific Acros
170430250 |
25 g | Glass bottle |
£276.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
617-90-3 | |
93.09 | |
YXDXXGXWFJCXEB-UHFFFAOYSA-N | |
69245 | |
N#CC1=CC=CO1 |
C5H3NO | |
MFCD00003223 | |
2-furonitrile, 2-cyanofuran, 2-furancarbonitrile, 2-furyl cyanide, unii-2lrk86h722, .alpha.-furyl cyanide, pubchem6961, acmc-209mwb, 2-cyanofuran;2-furancarbonitrile | |
furan-2-carbonitrile |
Specifications
617-90-3 | |
100.0 | |
1.0600g/mL | |
35°C | |
98.5% min. (GC) | |
1.4780 to 1.4800 | |
18, 278 | |
2-furonitrile, 2-cyanofuran, 2-furancarbonitrile, 2-furyl cyanide, unii-2lrk86h722, .alpha.-furyl cyanide, pubchem6961, acmc-209mwb, 2-cyanofuran;2-furancarbonitrile | |
YXDXXGXWFJCXEB-UHFFFAOYSA-N | |
furan-2-carbonitrile | |
69245 | |
99% | |
2-Furonitrile, 99% |
98.5 | |
Yellow to Orange | |
146.0°C to 148.0°C | |
Authentic | |
C5H3NO | |
MFCD00003223 | |
1.06 | |
Solubility in water: slightly. Other solubilities: soluble in most common organic solvents | |
N#CC1=CC=CO1 | |
93.09 | |
93.08 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-537-4
missing translation for 'tsca' : TSCA
Recommended Storage : Product may darken on storage
- 2-Furonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only