Learn More
2-Furoyl chloride, 98+%, Thermo Scientific Chemicals
£44.12 - £179.90
Chemical Identifiers
CAS | 527-69-5 |
---|---|
Molecular Formula | C5H3ClO2 |
Molecular Weight (g/mol) | 130.527 |
MDL Number | MFCD00003228 |
InChI Key | OFTKFKYVSBNYEC-UHFFFAOYSA-N |
Synonym | 2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride |
PubChem CID | 68242 |
IUPAC Name | furan-2-carbonyl chloride |
SMILES | C1=COC(=C1)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397066
|
Thermo Scientific Alfa Aesar
L06960.18 |
50 g |
£44.12
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307076
|
Thermo Scientific Alfa Aesar
L06960.30 |
250 g |
£179.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
527-69-5 | |
130.527 | |
OFTKFKYVSBNYEC-UHFFFAOYSA-N | |
68242 | |
C1=COC(=C1)C(=O)Cl |
C5H3ClO2 | |
MFCD00003228 | |
2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride | |
furan-2-carbonyl chloride |
Specifications
527-69-5 | |
1.325 | |
85°C (185°F) | |
1.531 | |
UN3265 | |
Moisture sensitive | |
2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride | |
C1=COC(=C1)C(=O)Cl | |
130.527 | |
130.53 | |
2-Furoyl chloride |
-2°C to +1°C | |
173°C to 174°C | |
C5H3ClO2 | |
MFCD00003228 | |
110144 | |
14,4310 | |
OFTKFKYVSBNYEC-UHFFFAOYSA-N | |
furan-2-carbonyl chloride | |
68242 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 208-422-9
missing translation for 'rtecsNumber' : LT9925000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 2-Furoyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 2-Furancarbonyl chloride
RUO – Research Use Only