Learn More
2-Furoyl chloride, 98+%, Thermo Scientific Chemicals
£24.00 - £275.04
Chemical Identifiers
CAS | 527-69-5 |
---|---|
Molecular Formula | C5H3ClO2 |
Molecular Weight (g/mol) | 130.53 |
MDL Number | MFCD00003228 |
InChI Key | OFTKFKYVSBNYEC-UHFFFAOYSA-N |
Synonym | 2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride |
PubChem CID | 68242 |
IUPAC Name | furan-2-carbonyl chloride |
SMILES | C1=COC(=C1)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10046243
|
Thermo Scientific Acros
165380250 |
25 mL | Glass bottle |
£24.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10478910
|
Thermo Scientific Acros
165381000 |
100 mL | Glass bottle |
£65.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337490
|
Thermo Scientific Acros
165385000 |
500 mL | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
527-69-5 | |
130.53 | |
OFTKFKYVSBNYEC-UHFFFAOYSA-N | |
68242 | |
C1=COC(=C1)C(=O)Cl |
C5H3ClO2 | |
MFCD00003228 | |
2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride | |
furan-2-carbonyl chloride |
Specifications
527-69-5 | |
100.0 | |
Yellow to Brown | |
173°C to 174°C | |
Authentic | |
C5H3ClO2 | |
MFCD00003228 | |
15, 4339 | |
2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride | |
OFTKFKYVSBNYEC-UHFFFAOYSA-N | |
furan-2-carbonyl chloride | |
68242 | |
98+% | |
2-Furoyl chloride, 98+% |
98.0 | |
-2°C | |
1.3200g/mL | |
85°C | |
98% min. (GC) | |
1.5300 to 1.532 | |
18, 276 | |
1.32 | |
Solubility in water: decomposes. Other solubilities: soluble in ether,toluene,methylene chloride,,dichloroethane,chlorobenzene,ethyl acetate,thf,and chloroform,decomposes in alcohol | |
C1=COC(=C1)C(=O)Cl | |
130.53 | |
130.53 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-422-9
missing translation for 'tsca' : TSCA
- 2-Furoyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only