missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Heptanone, 99%, Honeywell™
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Density | 0.82g/mL at 25°C |
Packaging | Glass bottle |
Linear Formula | CH3(CH2)4COCH3 |
Quantity | 1 L |
UN Number | UN1110 |
Beilstein | 1699063 |
Formula Weight | 114.19g/mol |
Percent Purity | 99% |
Chemical Name or Material | 2-Heptanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'