Learn More
Fisherbrand™ 2 Hole Natural Rubber Stopper
Prevents liquid chemical leaks or escape from container
Brand:  Fisherbrand™ RB105/2H
 View more versions of this product
Product Code. 11597642
  • £37.40 / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
46/55 IRHD
1/Ea.
105 mm
Stopper (2 holes)
54mm
Red
Natural Rubber