missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-1-naphthaldehyde, 98%

Product Code. 11446826
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11446826

Brand: Thermo Scientific Alfa Aesar B23951.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 708-06-5 | C11H8O2 | 172.183 g/mol

2-Hydroxy-1-naphthaldehyde is used in Merrified-type synthesis to deterime free amino acid groups in polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-1-naphthaldehyde is used in Merrified-type synthesis to deterime free amino acid groups in polymers.

Solubility
Soluble in water (40 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Store under dry inert gas. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 708-06-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00004005
InChI Key NTCCNERMXRIPTR-UHFFFAOYSA-N
Synonym 2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
PubChem CID 12819
IUPAC Name 2-hydroxynaphthalene-1-carbaldehyde
SMILES C1=CC=C2C(=C1)C=CC(=C2C=O)O

Specifications

Melting Point 77°C to 80°C
Boiling Point 191°C to 193°C (27 mmHg)
Odor Odorless
Quantity 100 g
UN Number UN3077
Sensitivity Air Sensitive
Solubility Information Soluble in water (40g/L at 20°C).
Formula Weight 172.18
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-1-naphthaldehyde
compliance-icons
Product Identifier
  • 2-Hydroxy-1-naphthaldehyde
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : 2-Hydroxy-1-naphthaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.