missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4,6-dimethoxybenzoic acid, 97%, Thermo Scientific™

Product Code. 11374584
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Hydroxy-4,6-dimethoxybenzoic acid is a reagent used in the synthesis of rac Zearalenone-d6. rac Zearalenone-d6 s an estrogenic mycotoxin produced by Fusarium fungi commonly found in grains. One of a group of compounds known as resorcylic acid lactones.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3187-19-7
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00013977
InChI Key LEIJLTBRQHFMDL-UHFFFAOYSA-N
Synonym 4,6-dimethoxysalicylic acid, 4,6-dimethoxy-2-hydroxybenzoic acid, benzoic acid, 2-hydroxy-4,6-dimethoxy, acmc-1covk, 2-hydroxy-4,6-dimethoxybenzoicacid, 2,4-dimethoxy-6-hydroxybenzoic acid, 2-hydroxy-4,6-dimethoxy benzoic acid, 2-hydroxy-4,6-dimethoxy-benzoic acid, benzoic acid,2-hydroxy-4,6-dimethoxy
PubChem CID 76656
IUPAC Name 2-hydroxy-4,6-dimethoxybenzoic acid
SMILES COC1=CC(=C(C(=C1)OC)C(=O)O)O

Specifications

Melting Point 160°C to 162°C
Quantity 5 g
Beilstein 2213838
Solubility Information Insoluble in water.
Formula Weight 198.17
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-4,6-dimethoxybenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'