missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.
Solubility
Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.
Solubility
Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 77°C to 79°C |
Density | 1.18 |
Quantity | 5 g |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 282.3 |
Percent Purity | 99% |
Chemical Name or Material | 2-Hydroxy-4-(methacryloyloxy)benzophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'