missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-n-octyloxybenzophenone, 99%, Thermo Scientific™

Product Code. 11342607
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1843-05-6
Molecular Formula C21H26O3
Molecular Weight (g/mol) 326.436
MDL Number MFCD00027327
InChI Key QUAMTGJKVDWJEQ-UHFFFAOYSA-N
Synonym octabenzone, 2-hydroxy-4-n-octyloxybenzophenone, benzophenone-12, 2-hydroxy-4-octyloxy phenyl phenyl methanone, benzon oo, 2-hydroxy-4-octyloxy benzophenone, benzophenone 12, cyasorb uv 531, spectra-sorb uv 531, sumisorb 130
PubChem CID 15797
IUPAC Name (2-hydroxy-4-octoxyphenyl)-phenylmethanone
SMILES CCCCCCCCOC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

Melting Point 46°C to 49°C
Quantity 25 g
Beilstein 1915198
Merck Index 14,6742
Formula Weight 326.44
Percent Purity 99%
Chemical Name or Material 2-Hydroxy-4-n-octyloxybenzophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'