missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-iodopyridine, 97%

Product Code. p-7047979
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11384456

Brand: Thermo Scientific Alfa Aesar L20044.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13472-79-2 | C5H4INO | 220.997 g/mol

Employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-79-2
Molecular Formula C5H4INO
Molecular Weight (g/mol) 220.997
MDL Number MFCD00234058
InChI Key ZDJUNNCVIDKJAN-UHFFFAOYSA-N
Synonym 2-hydroxy-5-iodopyridine, 5-iodopyridin-2-ol, 5-iodopyridin-2 1h-one, 5-iodo-pyridin-2-ol, 5-iodo-2-pyridone, 2 1h-pyridinone, 5-iodo, 5-iodo-2-pyridinol, 5-jod-2-pyridon, 5-iodo-2-pyridol, 5-iodopyridin-2-one
PubChem CID 459500
IUPAC Name 5-iodo-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1I

Specifications

Melting Point 189°C to 191°C
Quantity 1 g
Beilstein 108752
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 221
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-5-iodopyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.