missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
28.54 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Use promo code "21640" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
Description
2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.
Solubility
Miscible with chloroform. Slightly miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.
2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.
Solubility
Miscible with chloroform. Slightly miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 13°C to 16°C |
Density | 1.221 |
Boiling Point | 248°C to 252°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.578 |
Quantity | 25 g |
Beilstein | 1100532 |
Sensitivity | Air Sensitive |
Solubility Information | Miscible with chloroform. Slightly miscible with water. |
Formula Weight | 152.15 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'