missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11328696
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Hydroxybenzeneboronic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzeneboronic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89466-08-0
Molecular Formula C6H7BO3
Molecular Weight (g/mol) 137.93
MDL Number MFCD01074581
InChI Key YDMRDHQUQIVWBE-UHFFFAOYSA-N
Synonym 2-hydroxyphenyl boronic acid, 2-hydroxybenzeneboronic acid, o-hydroxyphenylboronic acid, 2-boronophenol, 2-hydroxyphenyl boranediol, 2-hydroxyphenylboronicacid, boronic acid, 2-hydroxyphenyl, pubchem9522, 2-hydroxyphenylboronic, acmc-209r1d
PubChem CID 2773454
IUPAC Name (2-hydroxyphenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1O

Specifications

Melting Point ∼185°C
Quantity 5 g
Beilstein 2962938
Solubility Information Insoluble in water.
Formula Weight 137.93
Percent Purity 97%
Chemical Name or Material 2-Hydroxybenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'