Learn More
2-Hydroxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
£44.10 - £169.00
Chemical Identifiers
CAS | 89466-08-0 |
---|---|
Molecular Formula | C6H7BO3 |
Molecular Weight (g/mol) | 137.93 |
MDL Number | MFCD01074581 |
InChI Key | YDMRDHQUQIVWBE-UHFFFAOYSA-N |
Synonym | 2-hydroxyphenyl boronic acid, 2-hydroxybenzeneboronic acid, o-hydroxyphenylboronic acid, 2-boronophenol, 2-hydroxyphenyl boranediol, 2-hydroxyphenylboronicacid, boronic acid, 2-hydroxyphenyl, pubchem9522, 2-hydroxyphenylboronic, acmc-209r1d |
PubChem CID | 2773454 |
IUPAC Name | (2-hydroxyphenyl)boronic acid |
SMILES | OB(O)C1=CC=CC=C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11318696
|
Thermo Scientific Alfa Aesar
L19400.03 |
1 g |
£44.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328696
|
Thermo Scientific Alfa Aesar
L19400.06 |
5 g |
£169.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Hydroxybenzeneboronic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxybenzeneboronic acid is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
89466-08-0 | |
137.93 | |
YDMRDHQUQIVWBE-UHFFFAOYSA-N | |
2773454 | |
OB(O)C1=CC=CC=C1O |
C6H7BO3 | |
MFCD01074581 | |
2-hydroxyphenyl boronic acid, 2-hydroxybenzeneboronic acid, o-hydroxyphenylboronic acid, 2-boronophenol, 2-hydroxyphenyl boranediol, 2-hydroxyphenylboronicacid, boronic acid, 2-hydroxyphenyl, pubchem9522, 2-hydroxyphenylboronic, acmc-209r1d | |
(2-hydroxyphenyl)boronic acid |
Specifications
89466-08-0 | |
C6H7BO3 | |
2962938 | |
Insoluble in water. | |
OB(O)C1=CC=CC=C1O | |
137.93 | |
137.93 | |
2-Hydroxybenzeneboronic acid |
∼185°C | |
MFCD01074581 | |
2-hydroxyphenyl boronic acid, 2-hydroxybenzeneboronic acid, o-hydroxyphenylboronic acid, 2-boronophenol, 2-hydroxyphenyl boranediol, 2-hydroxyphenylboronicacid, boronic acid, 2-hydroxyphenyl, pubchem9522, 2-hydroxyphenylboronic, acmc-209r1d | |
YDMRDHQUQIVWBE-UHFFFAOYSA-N | |
(2-hydroxyphenyl)boronic acid | |
2773454 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only