missing translation for 'onlineSavingsMsg'
Learn More
2-Hydroxychalcone, 98+%, Thermo Scientific Chemicals Product Code.: 11415263

2-Hydroxychalcone, 98+%, Thermo Scientific Chemicals

Product Code. 11415263
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11415263

Brand: Thermo Scientific Alfa Aesar A12851.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 644-78-0
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00016449
InChI Key UDOOPSJCRMKSGL-ZHACJKMWSA-N
Synonym 2-hydroxychalcone, 2e-3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 2-propen-1-one, 3-2-hydroxyphenyl-1-phenyl, 2-2-hydroxybenzal acetophenone, e-3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 3-2-hydroxyphenyl-1-phenylprop-2-en-1-one, 3-2-hydroxyphenyl-1-phenyl-2-propen-1-one, e-3-2-hydroxyphenyl-1-phenyl-prop-2-en-1-one, 2-hydroxybenzylidene acetophenone, chalcone, 1Show MoreShow Less
PubChem CID 5367146
IUPAC Name (E)-3-(2-hydroxyphenyl)-1-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2O

Specifications

Melting Point ∼154°C (decomposition)
Quantity 25 g
Beilstein 2049134
Formula Weight 224.26
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxychalcone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number90008717 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12851.06,A12851.14
Lot Number10221113 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12851.06,A12851.14
Lot Number10234781 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12851.06,A12851.14
Lot NumberE2766B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12851.06,A12851.14
Lot Number10203433 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12851.06,A12851.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Hydroxychalcone, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.