missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl methacrylate, 97%, stabilized

Product Code. 10488383 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
500g
This item is not returnable. View return policy

Product Code. 10488383

Brand: Thermo Scientific Acros 156332500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 868-77-9 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-77-9, 150-76-5
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00002863
InChI Key WOBHKFSMXKNTIM-UHFFFAOYSA-N
Synonym 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
PubChem CID 13360
ChEBI CHEBI:34288
IUPAC Name 2-hydroxyethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCO

Specifications

Acidity 2mg KOH/g max.
CAS Min % 96.0
CAS Max % 100.0
Melting Point -12.0°C
Density 1.0730g/mL
Boiling Point 213°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CH2OH
Refractive Index 1.4510 to 1.4530
Quantity 250 g
Beilstein 02,III,1291
Specific Gravity 1.073
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Stabilizer 200 to 400ppm MEHQ
Viscosity 5 mPa.s (30°C)
Formula Weight 130.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Hydroxyethyl methacrylate
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Hydroxyethyl methacrylate, 4-Methoxyphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.