missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Descripción
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.
Solubility
Soluble in Methanol and Water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.
Solubility
Soluble in Methanol and Water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Especificaciones
Especificaciones
Melting Point | 145°C to 147°C |
Quantity | 50 g |
Beilstein | 908000 |
Solubility Information | Soluble in Methanol and Water. |
Formula Weight | 152.15 |
Percent Purity | 98% |
Chemical Name or Material | 2-Hydroxyphenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'