missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11945861
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89466-08-0
Molecular Formula C6H7BO3
Molecular Weight (g/mol) 137.93
MDL Number MFCD01074581
InChI Key YDMRDHQUQIVWBE-UHFFFAOYSA-N
Synonym 2-hydroxyphenyl boronic acid, 2-hydroxybenzeneboronic acid, o-hydroxyphenylboronic acid, 2-boronophenol, 2-hydroxyphenyl boranediol, 2-hydroxyphenylboronicacid, boronic acid, 2-hydroxyphenyl, pubchem9522, 2-hydroxyphenylboronic, acmc-209r1d
PubChem CID 2773454
IUPAC Name (2-hydroxyphenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1O

Specifications

Melting Point 173.0°C to 174.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Linear Formula HOC6H4B(OH)2
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 137.93
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Hydroxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.