missing translation for 'onlineSavingsMsg'
Learn More
(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480, Thermo Scientific Chemicals Product Code.: 15437795

(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480, Thermo Scientific Chemicals

Product Code. 15437795
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15437795

Brand: Thermo Scientific Alfa Aesar H31133.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128446-35-5
Molecular Formula C44H75O36
Molecular Weight (g/mol) 1180.05
MDL Number MFCD00069372

Specifications

Melting Point 305°C
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight ≈1250 to 1480 d
Chemical Name or Material (2-Hydroxypropyl)-beta-cyclodextrin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247835 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberH31133.22,H31133.14,H31133.06
Lot Number10240159 Certificate TypeCertificate of Analysis Date13/04/2022 Catalog NumberH31133.22,H31133.14,H31133.06
Lot Number10170720 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31133.22,H31133.14,H31133.06
Lot Number10226486 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31133.22,H31133.14,H31133.06
Lot Number10208525 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31133.22,H31133.14,H31133.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.