missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxypropyl-β-cyclodextrin, Tocris Bioscience™
Click to view available options
Quantity:
1 g
Description
The most widely used modified cyclodextrin, the lipophilic cavity formed by 7 glucose units. Drug solubility in water is greatly enhanced by complexing with HBC.
Specifications
CAS | 128446-35-5 |
Molecular Formula | C44H75O36 |
Quantity | 1 g |
Molecular Weight (g/mol) | 1180.05 |
Formula Weight | 1400 |
Percent Purity | >98% |
Chemical Name or Material | 2-Hydroxypropyl-β-cyclodextrin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'