missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxypyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
Quantity | 5 g |
Beilstein | 115859 |
IUPAC Name | 2-oxo-1H-pyridine-4-carboxylic acid |
Formula Weight | 139.11 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 2-Hydroxypyridine-4-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'