Learn More
2-Hydroxyquinoline, 98%, Thermo Scientific Chemicals
£28.00 - £453.93
Chemical Identifiers
CAS | 59-31-4 |
---|---|
Molecular Formula | C9H7NO |
Molecular Weight (g/mol) | 145.16 |
MDL Number | MFCD00006743 |
InChI Key | LISFMEBWQUVKPJ-UHFFFAOYSA-N |
Synonym | 2-hydroxyquinoline, 2-quinolinol, quinolin-2-ol, carbostyril, 2 1h-quinolinone, 2-quinolone, quinolin-2 1h-one, quinolinol, quinolinone, alpha-quinolone |
PubChem CID | 6038 |
ChEBI | CHEBI:18289 |
IUPAC Name | 1H-quinolin-2-one |
SMILES | C1=CC=C2C(=C1)C=CC(=O)N2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10591124
|
Thermo Scientific Acros
418810010 |
1 g | Glass bottle |
£28.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10463361
|
Thermo Scientific Acros
418810100 |
10 g | Glass bottle | ||||||
10187750
|
Thermo Scientific Acros
418810250 |
25 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59-31-4 | |
145.16 | |
LISFMEBWQUVKPJ-UHFFFAOYSA-N | |
6038 | |
1H-quinolin-2-one |
C9H7NO | |
MFCD00006743 | |
2-hydroxyquinoline, 2-quinolinol, quinolin-2-ol, carbostyril, 2 1h-quinolinone, 2-quinolone, quinolin-2 1h-one, quinolinol, quinolinone, alpha-quinolone | |
CHEBI:18289 | |
C1=CC=C2C(=C1)C=CC(=O)N2 |
Specifications
59-31-4 | |
100.0 | |
Brown-Purple or Purple to White | |
97.5% min. (HPLC) | |
MFCD00006743 | |
2-hydroxyquinoline, 2-quinolinol, quinolin-2-ol, carbostyril, 2 1h-quinolinone, 2-quinolone, quinolin-2 1h-one, quinolinol, quinolinone, alpha-quinolone | |
C1=CC=C2C(=C1)C=CC(=O)N2 | |
145.16 | |
CHEBI:18289 | |
98% | |
2-Hydroxyquinoline, 98% |
98.5 | |
197.0°C to 200.0°C | |
Authentic | |
C9H7NO | |
15, 1826 | |
LISFMEBWQUVKPJ-UHFFFAOYSA-N | |
1H-quinolin-2-one | |
6038 | |
145.16 | |
Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-420-6
missing translation for 'rtecsNumber' : FG7175000
missing translation for 'tsca' : TSCA
- 2-Hydroxyquinoline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only