missing translation for 'onlineSavingsMsg'
Learn More

2-Indanone, 97%, Thermo Scientific Chemicals

Product Code. 10604842
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-13-4
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00003792
InChI Key UMJJFEIKYGFCAT-UHFFFAOYSA-N
Synonym 2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone
PubChem CID 11983
ChEBI CHEBI:27930
IUPAC Name 1,3-dihydroinden-2-one
SMILES C1C(=O)CC2=CC=CC=C21

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50.0°C to 57.0°C
Color Brown-Yellow to Yellow
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 07, 363
Solubility Information Solubility in water: insoluble
Formula Weight 132.16
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 2-Indanone, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'