missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Indanone, 97%, Thermo Scientific Chemicals
£49.90 - £225.00
Chemical Identifiers
CAS | 615-13-4 |
---|---|
Molecular Formula | C9H8O |
Molecular Weight (g/mol) | 132.16 |
MDL Number | MFCD00003792 |
InChI Key | UMJJFEIKYGFCAT-UHFFFAOYSA-N |
Synonym | 2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone |
PubChem CID | 11983 |
ChEBI | CHEBI:27930 |
IUPAC Name | 1,3-dihydroinden-2-one |
SMILES | C1C(=O)CC2=CC=CC=C21 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10604842
|
Thermo Scientific Acros
164110100 |
10 g | Glass bottle |
£49.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10731631
|
Thermo Scientific Acros
164110500 |
50 g | Glass bottle |
£225.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
615-13-4 | |
132.16 | |
UMJJFEIKYGFCAT-UHFFFAOYSA-N | |
11983 | |
1,3-dihydroinden-2-one |
C9H8O | |
MFCD00003792 | |
2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone | |
CHEBI:27930 | |
C1C(=O)CC2=CC=CC=C21 |
Specifications
615-13-4 | |
100.0 | |
Brown-Yellow to Yellow | |
Authentic | |
C9H8O | |
07, 363 | |
Solubility in water: insoluble | |
C1C(=O)CC2=CC=CC=C21 | |
132.16 | |
CHEBI:27930 | |
97% | |
2-Indanone, 97% |
96.0 | |
50.0°C to 57.0°C | |
100°C | |
96% min. (GC) | |
MFCD00003792 | |
2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone | |
UMJJFEIKYGFCAT-UHFFFAOYSA-N | |
1,3-dihydroinden-2-one | |
11983 | |
132.16 | |
Crystals or Powder |
Safety and Handling
missing translation for 'einecsNumber' : 210-410-3
missing translation for 'rtecsNumber' : NK7535500
missing translation for 'tsca' : TSCA
RUO – Research Use Only