missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-1,3,5-trimethylbenzene, 98%, Thermo Scientific Chemicals

Product Code. 11428296
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4028-63-1
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.091
MDL Number MFCD00013707
InChI Key GTPNXFKONRIHRW-UHFFFAOYSA-N
Synonym 2,4,6-trimethyliodobenzene, 2-iodomesitylene, mesityl iodide, benzene, 2-iodo-1,3,5-trimethyl, iodo-2,4,6-trimethylbenzene, 2,4,6-trimethyliodobenzene 2-iodomesitylene, iodomesitylene, jodmesitylen, mesityliodine, pubchem12601
PubChem CID 77647
IUPAC Name 2-iodo-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)I)C

Specifications

Melting Point 28°C to 30°C
Quantity 100 g
Beilstein 1860064
Sensitivity Light sensitive
Formula Weight 246.09
Percent Purity 98%
Chemical Name or Material 2-Iodo-1,3,5-trimethylbenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'