missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5-nitrotoluene, 98%, Thermo Scientific™

Product Code. 11372067
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5326-38-5
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.03
MDL Number MFCD00051517
InChI Key ARJHCXYRCLMLQN-UHFFFAOYSA-N
Synonym 2-iodo-5-nitrotoluene, benzene, 1-iodo-2-methyl-4-nitro, 1-iodo-2-methyl-4-nitro-benzene, acmc-1amrp, 2-jod-5-nitro-1-methylbenzol, benzene,1-iodo-2-methyl-4-nitro
PubChem CID 79225
IUPAC Name 1-iodo-2-methyl-4-nitrobenzene
SMILES CC1=CC(=CC=C1I)[N+]([O-])=O

Specifications

Melting Point 99°C to 104°C
Quantity 5 g
Beilstein 2438587
Sensitivity Light sensitive
Formula Weight 263.03
Percent Purity 98%
Chemical Name or Material 2-Iodo-5-nitrotoluene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'