missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodoethanol, 99%, stab. with copper, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.
Solubility
Soluble in water.
Notes
Light sensitive. Store in a cool place. Incompatible with acid anhydrides, acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.
2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.
Solubility
Soluble in water.
Notes
Light sensitive. Store in a cool place. Incompatible with acid anhydrides, acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 2.202 |
Boiling Point | 85°C (25 mmHg) |
Flash Point | 65°C (149°F) |
Odor | Characteristic |
Linear Formula | ICH2CH2OH |
Refractive Index | 1.572 |
Quantity | 5 g |
UN Number | UN2810 |
Beilstein | 1098258 |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'