missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenylacetic acid, 97+%, Thermo Scientific Chemicals

Product Code. 10529813
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18698-96-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.04
MDL Number MFCD00046546
InChI Key IUHXGZHKSYYDIL-UHFFFAOYSA-N
Synonym 2-iodophenylacetic acid, 2-2-iodophenyl acetic acid, benzeneacetic acid, 2-iodo, 2-iodophenyl acetic acid, o-iodophenylacetic acid, 2-iodobenzeneacetic acid, 2-iodophenylaceticacid, pubchem2552, acmc-209eox, maybridge1_005605
PubChem CID 2780090
IUPAC Name 2-(2-iodophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)I

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 112°C to 114°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Linear Formula IC6H4CH2CO2H
Quantity 1 g
Formula Weight 262.04
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 2-Iodophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'